Tisk…

Chloroacetyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

259322-1G 259322-5G
SIAL259322-1GEA 1210 CZK
SIAL259322-1G SIAL259322-5G
Chloroacetyl isocyanate, Sigma-Aldrich®
Chloroacetyl isocyanate
Vzorec: C₃H₂ClNO₂
MDL Number: MFCD00002034
CAS číslo: 4461-30-7
EINECS: 224-715-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR