Tisk…

DL-Malic acid monosodium salt ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A19428.18 A19428.30
A19428.18EA 1490 CZK
A19428.18 A19428.30
DL-Malic acid monosodium salt ≥98%
DL-Malic acid monosodium salt

Purity: 98% CAS No.: 57467-17-1 Molecular Formula: C4H5NaO5 Formula Weight: 156.07 Melting Point: 210° dec. MDL No.: MFCD00798569

Vzorec: C₄H₅NaO₅
Storage Temperature: Ambient
MDL Number: MFCD00798569
CAS číslo: 57467-17-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR