Tisk…
Octyltrimethylammonium bromide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Trimethyl-1-octylammonium bromide
75091-25G 75091-5G
SIAL75091-25GEA 4890 CZK
SIAL75091-25G SIAL75091-5G
Octyltrimethylammonium bromide, Sigma-Aldrich®
Octyltrimethylammonium bromide
Vzorec: C₁₁H₂₆BrN
MDL Number: MFCD00051873
CAS číslo: 2083-68-3