Tisk…

Hexyltrimethylammonium bromide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hexyltrimethylammonium Bromide, (1-Hexyl)trimethylammonium bromide

L08594.06 L08594.14
L08594.06EA 1320 CZK
L08594.06 L08594.14
Hexyltrimethylammonium bromide ≥98%
Hexyltrimethylammonium bromide

Purity: 98% CAS No.: 2650-53-5 Molecular Formula: CH3(CH2)5N(CH3)3Br Formula Weight: 224.18 Melting Point: 177-180° Beilstein Reference: 3913229

Vzorec: C₉H₂₂BrN
Bod tání: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00051872
CAS číslo: 2650-53-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR