Tisk…
Ethanesulphonic acid ≥70%, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
M-Clarity™ quality level = MQ100
471550-100G 471550-500G
SIAL471550-100GEA 0 CZK
SIAL471550-100G SIAL471550-500G
Ethanesulphonic acid ≥70%, Sigma-Aldrich®
Ethanesulphonic acid
Vzorec: C₂H₆O₃S
MW: 110,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007529
CAS číslo: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8,III