Tisk…

Di-tert-butyl malonate 98 Stabilised

Dodavatel: Thermo Fisher Scientific
Varování


Stabilization: K2CO3

A12774.09 A12774.18 A12774.30
A12774.09EA 1690 CZK
A12774.09 A12774.18 A12774.30
Di-tert-butyl malonate 98 Stabilised
Di-tert-butyl malonate

Stabilized with potassium carbonate


Purity: 98+% CAS No.: 541-16-2 Molecular Formula: C11H20O4 Formula Weight: 216.28 Flash Point: 88°(190°F) Refractive Index: 1.4190 Melting Point: -6° Boiling Point: 65-67°/1mm Beilstein Reference: 1781766 Merck Reference: 14,3034 MDL No.: MFCD00008810

Vzorec: C₁₁H₂₀O₄
MW: 216.28 g/mol
Bod varu: 65…67 °C (1 mmHg)
Bod tání: –6 °C
Hustota: 0.965
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008810
CAS číslo: 541-16-2
EINECS: 208-769-6
Merck Index: 14,3034

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR