tert-Butyl methyl malonate 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
tert.-Butyl methyl malonate
339390050
339390250
ACRO339390050EA
1320
CZK
ACRO339390050
ACRO339390250
tert-Butyl methyl malonate 95%
tert-Butyl methyl malonate
Vzorec:
C₈H₁₄O₄ Bod varu: 80 °C (11 mmHg) Bod tání: –10 °C Hustota: 1,03 g/cm³ Bod vzplanutí: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042856 CAS číslo: 42726-73-8 EINECS: 255-919-1 |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid (May discolor to yellow on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4140 to 1.4170 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...