Tisk…

4,6-Dichlororesorcinol ≥97%

Dodavatel: abcr
Varování

AB126690.0025 AB126690.0100 AB126690.0500
ABCRAB126690.0025EA 0 CZK
ABCRAB126690.0025 ABCRAB126690.0100 ABCRAB126690.0500
4,6-Dichlororesorcinol ≥97%
4,6-Dichlororesorcinol

Purity: 97% CAS No.: 137-19-9 Molecular Formula: C6H4Cl2O2 Formula Weight: 179.00 Melting Point: 113-116° Boiling Point: 253-255° MDL No.: MFCD00002274

Vzorec: C₆H₄Cl₂O₂
MW: 179.00 g/mol
Bod varu: 253…255 °C
Bod tání: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002274
CAS číslo: 137-19-9
EINECS: 205-284-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR