Tisk…

1,4,7-Triazacyclononane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4,7-Triazonane, TACN

M-Clarity™ quality level = MQ200

311308-100MG 311308-500MG
SIAL311308-100MGEA 2420 CZK
SIAL311308-100MG SIAL311308-500MG
1,4,7-Triazacyclononane, Sigma-Aldrich®
1,4,7-Triazacyclononane
Vzorec: C₆H₁₅N₃
MW: 129,2 g/mol
MDL Number: MFCD00012358
CAS číslo: 4730-54-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR