2-Cyanopyrazine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
M-Clarity™ quality level = MQ100
349496-10G
349496-50G
SIAL349496-10GEA
1610
CZK
SIAL349496-10G
SIAL349496-50G
2-Cyanopyrazine, Sigma-Aldrich®
2-Cyanopyrazine
Vzorec:
C₅H₃N₃ |
MDL Number:
MFCD00049361 CAS číslo: 19847-12-2 EINECS: 243-369-5 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...