Tisk…

(S)-(+)-3-Hydroxytetrahydrofuran, Sigma-Aldrich®

Dodavatel: Merck


MQ100

296686-25G 296686-5G
SIAL296686-25GEA 15060 CZK
SIAL296686-25G SIAL296686-5G
(S)-(+)-3-Hydroxytetrahydrofuran, Sigma-Aldrich®
(S)-(+)-3-Hydroxytetrahydrofuran
Vzorec: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD00064327
CAS číslo: 86087-23-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR