Tisk…

Tetrabutylammonium azide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

651664-25G 651664-5G
SIAL651664-25GEA 11720 CZK
SIAL651664-25G SIAL651664-5G
Tetrabutylammonium azide, Sigma-Aldrich®
Tetrabutylammonium azide
Vzorec: C₁₆H₃₆N₄
MDL Number: MFCD00060069
CAS číslo: 993-22-6
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR