Tisk…
Tetrabutylammonium dihydrogen phosphate 0.5 M in HPLC grade water for HPLC
Dodavatel: Thermo Fisher Scientific
Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
420170250 420175000 420171000
ACRO420170250EA 1880 CZK
ACRO420170250 ACRO420175000 ACRO420171000
Tetrabutylammonium dihydrogen phosphate 0.5 M in HPLC grade water for HPLC
Tetrabutylammonium dihydrogen phosphate
Vzorec: C₁₆H₃₈NO₄P
MW: 339.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064526
CAS číslo: 5574-97-0
EINECS: 226-947-1
Objednat
Výsledky specifikačního testu
Appearance Clear solution
UV at 300 nm A: ≤0.03
Color scale ≤10 APHA
Titration with NaOH 0.48 to 0.52 M