Tisk…

Benzyltrimethylammonium dichloroiodate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

343366-25G 343366-5G
SIAL343366-25GEA 7400 CZK
SIAL343366-25G SIAL343366-5G
Benzyltrimethylammonium dichloroiodate, Sigma-Aldrich®
Benzyltrimethylammonium dichloroiodate
Vzorec: C₁₀H₁₆Cl₂IN
MW: 348,06 g/mol
MDL Number: MFCD00075259
CAS číslo: 114971-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR