Tisk…

Disodium mesoxalate monohydrate, Sigma-Aldrich®

Dodavatel: Merck

71740-10G 71740-50G
SIAL71740-10GEA 0 CZK
SIAL71740-10G SIAL71740-50G
Disodium mesoxalate monohydrate, Sigma-Aldrich®
Disodium mesoxalate monohydrate
Vzorec: C₃Na₂O₅·1H₂O
CAS číslo: 31635-99-1
EINECS: 230-867-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR