Tisk…

1,4,7-Trimethyl-1,4,7-triazacyclononane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4,7-Trimethyl-1,4,7-triazonane

M-Clarity™ quality level = MQ100

311294-500MG 311294-100MG
SIAL311294-500MGEA 7420 CZK
SIAL311294-500MG SIAL311294-100MG
1,4,7-Trimethyl-1,4,7-triazacyclononane, Sigma-Aldrich®
1,4,7-Trimethyl-1,4,7-triazacyclononane
Vzorec: C₉H₂₁N₃
MW: 171,28 g/mol
MDL Number: MFCD00012359
CAS číslo: 96556-05-7
UN: 3267
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR