Tisk…

4-Pentylcyclohexanone ≥98%

Dodavatel: Thermo Scientific

B22088.09 B22088.18
B22088.09EA 1640 CZK
B22088.09 B22088.18
4-Pentylcyclohexanone ≥98%
4-Pentylcyclohexanone

Purity: 98+% CAS No.: 61203-83-6 Molecular Formula: C11H20O Formula Weight: 168.28 UN No.: 3082 MDL No.: MFCD00059581

Vzorec: C₁₁H₂₀O
MW: 168,28 g/mol
Hustota: 0,89
Storage Temperature: Ambient
MDL Number: MFCD00059581
CAS číslo: 61203-83-6
EINECS: 406-670-4
UN: 3082
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR