2-Ethylcyclohexanone ≥99%
Dodavatel: Thermo Scientific
B20748.06
B20748.14
B20748.06EA
2630
CZK
B20748.06
B20748.14
2-Ethylcyclohexanone ≥99%
2-Ethylcyclohexanone
Purity: 99%
CAS No.: 4423-94-3
Molecular Formula: C8H14O
Formula Weight: 126.20
MDL No.: MFCD00066306
Vzorec:
C₈H₁₄O MW: 126,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066306 CAS číslo: 4423-94-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...