(4-Nitrophenyl)-α-D-glucopyranoside 99%
Dodavatel: Thermo Fisher Scientific
337150050
337150010
337150250
ACRO337150050EA
9200
CZK
ACRO337150050
ACRO337150010
ACRO337150250
(4-Nitrophenyl)-α-D-glucopyranoside 99%
(4-Nitrophenyl)-α-D-glucopyranoside
Vzorec:
C₁₂H₁₅NO₈ Bod tání: 204…218 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064088 CAS číslo: 3767-28-0 EINECS: 223-189-3 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Solubility | (2% in methanol) Clear to slightly hazy colorless to green-yellow solution |
HPLC | ≥98.5 % |
Specific optical rotation | +210° to +220° (20°C, 589 nm) (c=1, water) |
Impurity | ≤0.1 % (p-Nitrophenol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...