Tisk…
Carmine alum lake of carminic acid, pure_certified
Dodavatel: Thermo Fisher Scientific
Synonyms: CI 75470
404140250 404141000
ACRO404140250EA 5340 CZK
ACRO404140250 ACRO404141000
Carmine alum lake of carminic acid, pure_certified
Carmine
Vzorec: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167028
CAS číslo: 1390-65-4
EINECS: 215-724-4
Objednat
Výsledky specifikačního testu
Appearance Dark red Powder
Infrared spectrum Conforms
UV spectrum Complies
BSC Certification Passes