Tisk…

2,2-Dimethylpropane-1,3-diamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,2-Dimethyl-1,3-propanediamine

M-Clarity™ quality level = MQ200

226920-250G 226920-50G
SIAL226920-250GEA 12280 CZK
SIAL226920-250G SIAL226920-50G
2,2-Dimethylpropane-1,3-diamine, Sigma-Aldrich®
2,2-Dimethylpropane-1,3-diamine
Vzorec: C₅H₁₄N₂
MW: 102,1800 g/mol
MDL Number: MFCD00009801
CAS číslo: 7328-91-8
EINECS: 230-819-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR