Phenylglyoxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Oxo-2-phenylacetic acid, Benzoylformic acid
M-Clarity™ quality level = MQ200
B13055-100G
B13055-25G
B13055-5G
SIALB13055-100GEA
14330
CZK
SIALB13055-100G
SIALB13055-25G
SIALB13055-5G
Phenylglyoxylic acid, Sigma-Aldrich®
Phenylglyoxylic acid
Vzorec:
C₈H₆O₃ MW: 150.13 g/mol |
MDL Number:
MFCD00002575 CAS číslo: 611-73-4 EINECS: 210-278-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...