Tisk…

2-(2-Bromoacetyl)thiophene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromo-1-(thiophen-2-yl)ethanone, 2-(Bromoacetyl)thiophene

M-Clarity™ quality level = MQ100

595594-1G 595594-5G
SIAL595594-1GEA 3360 CZK
SIAL595594-1G SIAL595594-5G
2-(2-Bromoacetyl)thiophene, Sigma-Aldrich®
2-(2-Bromoacetyl)thiophene
Vzorec: C₆H₅BrOS
MDL Number: MFCD02677721
CAS číslo: 10531-41-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR