Tisk…
Hydroxid sodný-D 40% in D₂O (99.5% D) (Isotopic), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
372072-10G 372072-50G
SIAL372072-10GEA 2380 CZK
SIAL372072-10G SIAL372072-50G
Hydroxid sodný-D 40% in D₂O (99.5% D) (Isotopic), Sigma-Aldrich®
Hydroxid sodný-D
Vzorec: NaOD
MW: 41.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037669
CAS číslo: 14014-06-3
EINECS: 237-825-2
UN: 1824
ADR: 8 II