Tisk…

2-Isopropyl-3-methoxypyrazine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

243132-1G 243132-5G
SIAL243132-1GEA 1560 CZK
SIAL243132-1G SIAL243132-5G
2-Isopropyl-3-methoxypyrazine, Sigma-Aldrich®
2-Isopropyl-3-methoxypyrazine
Vzorec: C₈H₁₂N₂O
MDL Number: MFCD00006135
CAS číslo: 25773-40-4
EINECS: 247-256-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR