Tisk…

Tetrabutylammonium cyanante, Sigma-Aldrich®

Dodavatel: Merck

86864-25G 86864-5G
SIAL86864-25GEA 12760 CZK
SIAL86864-25G SIAL86864-5G
Tetrabutylammonium cyanante, Sigma-Aldrich®
Tetrabutylammonium cyanante
Vzorec: C₁₇H₃₆N₂O
CAS číslo: 39139-87-2
EINECS: 254-315-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR