Ethyl phenylglyoxylate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethyl benzoylformate
M-Clarity™ quality level = MQ100
258911-25G
258911-5G
SIAL258911-25GEA
3050
CZK
SIAL258911-25G
SIAL258911-5G
Ethyl phenylglyoxylate, Sigma-Aldrich®
Ethyl phenylglyoxylate
Vzorec:
C₁₀H₁₀O₃ |
MDL Number:
MFCD00009120 CAS číslo: 1603-79-8 EINECS: 216-504-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...