Tisk…

Ethyl phenylglyoxylate ≥98%

Dodavatel: Thermo Fisher Scientific

B25292.06 B25292.14 B25292.22
B25292.06EA 944 CZK
B25292.06 B25292.14 B25292.22
Ethyl phenylglyoxylate ≥98%
Ethyl phenylglyoxylate

Purity: 98% CAS No.: 1603-79-8 Molecular Formula: C10H10O3 Formula Weight: 178.19 Flash Point: 110°(230°F) Refractive Index: 1.5160 Boiling Point: 138-139°/18mm MDL No.: MFCD00009120

Vzorec: C₁₀H₁₀O₃
Bod varu: 138…139 °C (18 mmHg)
Hustota: 1,122
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009120
CAS číslo: 1603-79-8
EINECS: 216-504-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR