Tisk…

Hexyl isovalerate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W349909-1KG-K W349909-9KG-K
SIALW349909-1KG-KEA 3900 CZK
SIALW349909-1KG-K SIALW349909-9KG-K
Hexyl isovalerate, Sigma-Aldrich®
Hexyl isovalerate
Vzorec: C₁₁H₂₂O₂
CAS číslo: 10032-13-0
EINECS: 233-105-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR