3,5-Difluoro-4-methoxyaniline ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H26778.03
H26778.06
H26778.03EA
5700
CZK
H26778.03
H26778.06
3,5-Difluoro-4-methoxyaniline ≥97%
3,5-Difluoro-4-methoxyaniline
Purity: 97%
CAS No.: 363-47-3
Molecular Formula: C7H7F2NO
Formula Weight: 159.14
Odour: Characteristic
Melting Point: 77-80°
UN No.: 2811
MDL No.: MFCD04115910
Vzorec:
C₇H₇F₂NO MW: 159,14 g/mol Bod tání: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115910 CAS číslo: 363-47-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...