3-Methyl-1,2-cyclopentanedione, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methyl-1,2-cyclopentanedione, Maple lactone, Methyl cyclopentenolone, 3-Methylcyclopentane-1,2-dione
M-Clarity™ quality level = MQ300
W270008-1KG-K
W270008-25KG-K
W270008-5KG-K
SIALW270008-1KG-KEA
5450
CZK
SIALW270008-1KG-K
SIALW270008-25KG-K
SIALW270008-5KG-K
3-Methyl-1,2-cyclopentanedione, Sigma-Aldrich®
3-Methyl-1,2-cyclopentanedione
Vzorec:
C₆H₈O₂ MW: 112,13 g/mol Bod varu: 210 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00001417 CAS číslo: 765-70-8 EINECS: 212-154-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...