Tisk…

3-Methylbenzamidoxime

Dodavatel: Cayman Chemical

Synonyms: N-Hydroxy-3-methylbenzimidamide

13057-500 13057-1 13057-5
CAYM13057-500EA 1540 CZK
CAYM13057-500 CAYM13057-1 CAYM13057-5
3-Methylbenzamidoxime
3-Methylbenzamidoxime
Vzorec: C₈H₁₀N₂O
MW: 150,18 g/mol
MDL Number: MFCD00833266
CAS číslo: 40067-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR