4-Methoxythiobenzamide
Dodavatel: Cayman Chemical
Synonyms:
4-Methoxybenzothioamide
10552-500
10552-1
10552-5
CAYM10552-500EA
858
CZK
CAYM10552-500
CAYM10552-1
CAYM10552-5
4-Methoxythiobenzamide
4-Methoxythiobenzamide
Vzorec:
C₈H₉NOS Bod varu: 282 °C (1013 hPa) Bod tání: 155 °C |
MDL Number:
MFCD00040993 CAS číslo: 2362-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...