Tisk…

Dibenzosuberenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one, 5H-Dibenzo[a,d][7]annulen-5-one

D31737-100G D31737-25G
SIALD31737-100GEA 5350 CZK
SIALD31737-100G SIALD31737-25G
Dibenzosuberenone, Sigma-Aldrich®
Dibenzosuberenone
Vzorec: C₁₅H₁₀O
MW: 206,24 g/mol
Bod varu: 345 °C (1013 hPa)
Bod tání: 195…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00003588
CAS číslo: 2222-33-5
EINECS: 218-737-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR