Tisk…

Pyridoxal-5'-phosphate monohydrate ~98%, off-white powder

Dodavatel: MP Biomedicals

0210276925 0210276901 0210276905
ICNA0210276925EA 13980 CZK
ICNA0210276925 ICNA0210276901 ICNA0210276905
Pyridoxal-5'-phosphate monohydrate ~98%, off-white powder
Pyridoxal-5'-phosphate monohydrate

Pyridoxal-5-Phosphate Monohydrate is an active form of Vitamin B6, an enzyme prosthetic group.


Pyridoxal-5-Phosphate Monohydrate is useful for modifying lysyl and valyl residues in proteins. It acts as a coenzyme in transamination, decarboxylation and deamination reactions such as the conversion of dopa into dopamine; of glutamate into GABA; of histidine to histamine. PLP works mechanistically through formation of a Schiff-base.


Soluble in 1 M Hydrochloric acid (50 mg/ml - clear, colourless to light yellow-green solution).

Vzorec: C₈H₁₀NO₆P·1H₂O
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS číslo: 41468-25-1

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Moisture Content ~6.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR