Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine
A16606.06
A16606.14
A16606.06EA
2930
CZK
A16606.06
A16606.14
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Diphenylboric acid-2-aminoethyl ester (2-APB)
Purity: 98%
CAS No.: 524-95-8
Molecular Formula: C14H16BNO
Formula Weight: 225.10
Melting Point: 187-190°
Beilstein Reference: 2942273
Vzorec:
C₁₄H₁₆BNO MW: 225.1 g/mol Bod tání: 187…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014823 CAS číslo: 524-95-8 EINECS: 208-366-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...