Tisk…
Nalidixic acid ≥98%, high purity
Dodavatel: VWR Chemicals
Nebezpečí
0677-250GEA 12280 CZK
0677-250G
Nalidixic acid ≥98%, high purity
Nalidixic acid
A bactericidal agent that inhibits DNA gyrase activity. Recommended working concentration: 15 μg/ml.
Vzorec: C₁₂H₁₂N₂O₃
MW: 232,24 g/mol
Bod tání: 227°C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS číslo: 389-08-2
EINECS: 206-864-7
Objednat
Výsledky specifikačního testu
Heavy Metals (as Pb) < = 0.002 %
Loss on Drying <= 0.5 %
Melting Point 225 °C - 231 °C
Purity >= 98 %
Residue on Ignition <= 0.2 %