Tisk…

2-(Chloromethyl)quinoline hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

C57103-25G C57103-5G
SIALC57103-25GEA 0 CZK
SIALC57103-25G SIALC57103-5G
2-(Chloromethyl)quinoline hydrochloride, Sigma-Aldrich®
2-(Chloromethyl)quinoline hydrochloride
Vzorec: C₁₀H₉Cl₂N
MW: 214,10 g/mol
MDL Number: MFCD00012734
CAS číslo: 3747-74-8
EINECS: 223-145-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR