2-(Chloromethyl)quinoline hydrochloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A14102.03
A14102.06
A14102.03EA
1010
CZK
A14102.03
A14102.06
2-(Chloromethyl)quinoline hydrochloride ≥97%
2-(Chloromethyl)quinoline hydrochloride
Purity: 97%
CAS No.: 3747-74-8
Molecular Formula: C10H8ClN.HCl
Formula Weight: 214.10
Melting Point: 184-190°
Beilstein Reference: 3565832
Vzorec:
C₁₀H₉Cl₂N MW: 214,10 g/mol Bod tání: 184…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012734 CAS číslo: 3747-74-8 EINECS: 223-145-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...