Riboflavine-5'-phosphate sodium salt, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium
M-Clarity™ quality level = MQ200
R7774-100G
R7774-10G
R7774-25G
SIALR7774-100GEA
37630
CZK
SIALR7774-100G
SIALR7774-10G
SIALR7774-25G
Riboflavine-5'-phosphate sodium salt, Sigma-Aldrich®
Riboflavine-5'-phosphate sodium salt
Vzorec:
C₁₇H₂₀N₄NaO₉P MW: 478.3256 g/mol |
MDL Number:
MFCD00065362 CAS číslo: 130-40-5 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...