Tisk…

Riboflavine-5'-phosphate sodium salt, Sigma-Aldrich®

Dodavatel: Merck

F2253-100MG F2253-10MG F2253-25MG
SIALF2253-100MGEA 7090 CZK
SIALF2253-100MG SIALF2253-10MG SIALF2253-25MG
Riboflavine-5'-phosphate sodium salt, Sigma-Aldrich®
Riboflavine-5'-phosphate sodium salt
Vzorec: C₁₇H₂₀N₄NaO₉P
MDL Number: MFCD00065362
CAS číslo: 130-40-5
EINECS: 204-988-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR