L(-)-Carnitine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
C0283-1G
C0283-100G
C0283-10MG
C0283-25G
C0283-5G
SIALC0283-1GEA
1770
CZK
SIALC0283-1G
SIALC0283-100G
SIALC0283-10MG
SIALC0283-25G
SIALC0283-5G
L(-)-Carnitine hydrochloride, Sigma-Aldrich®
L(-)-Carnitine hydrochloride
Vzorec:
C₇H₁₆ClNO₃ |
MDL Number:
MFCD00066100 CAS číslo: 6645-46-1 EINECS: 229-663-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...