Boron trifluoride acetic acid complex (1:2), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Boron fluoride, acetic acid complex, Boron trifluoride - acetic acid complex, Trifluoroborane compound with acetic acid (1:2)
M-Clarity™ quality level = MQ100
359955-1L
359955-250ML
359955-5ML
SIAL359955-1LEA
4130
CZK
SIAL359955-1L
SIAL359955-250ML
SIAL359955-5ML
Boron trifluoride acetic acid complex (1:2), Sigma-Aldrich®
Boron trifluoride acetic acid complex (1:2)
Vzorec:
C₄H₈BF₃O₄ MW: 187,91 g/mol |
MDL Number:
MFCD00036359 CAS číslo: 373-61-5 EINECS: 206-768-5 UN: 1742 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...