Milk, non fat (skimmed milk), powder, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M7409-1BTL
M7409-5BTL
SIALM7409-1BTLEA
1460
CZK
SIALM7409-1BTL
SIALM7409-5BTL
Milk, non fat (skimmed milk), powder, Sigma-Aldrich®
Milk, non fat (skim milk)
Storage Temperature:
Ambient |
CAS číslo:
999999-99-4 EINECS: 310-127-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...