Tisk…

Milk, non fat (skimmed milk), powder

Dodavatel: PanReac AppliChem

A0830.0500 A0830.1000 A0830.5000
A0830.0500EA 1030 CZK
A0830.0500 A0830.1000 A0830.5000
Milk, non fat (skimmed milk), powder
Milk, non fat (skim milk)
Storage Temperature: Ambient
CAS číslo: 999999-99-4
EINECS: 310-127-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR