Tisk…

Isophorondiamine for synthesis, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 1-Amino-3-aminomethyl-3,5,5-trimethylcyclohexane, Isophoronediamine

M-Clarity™ quality level = MQ200

8.14123.0100 8.14123.0500
8.14123.0100EA 969 CZK
8.14123.0100 8.14123.0500
Isophorondiamine for synthesis, Sigma-Aldrich®
Isophorondiamine
Vzorec: C₁₀H₂₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00019397
CAS číslo: 2855-13-2
EINECS: 220-666-8
UN: 2289
ADR: 8,III

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Melting Point 10 °C
Molar Mass 170.3 g/mol
Boiling Point 247 °C (1013 hPa)
Vapor Pressure 0.02 hPa (20 °C)
Flash Point 112 °C
Refractive Index 1.4888 (20 °C, 589 nm)
Explosion Limit 1.2 %(V)
Density 0.92 g/cm3 (20 °C)
pH 11.6 (10 g/l, H2O, 20 °C)
Ignition Point 380 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR