Tisk…

1,3-Dicyclohexylimidazolium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Dicyclohexylimidazolium tetrafluoroborate salt

M-Clarity™ quality level = MQ100

666181-1G 666181-250MG 666181-5G
SIAL666181-1GEA 3780 CZK
SIAL666181-1G SIAL666181-250MG SIAL666181-5G
1,3-Dicyclohexylimidazolium tetrafluoroborate, Sigma-Aldrich®
1,3-Dicyclohexylimidazolium tetrafluoroborate
Vzorec: C₁₅H₂₅BF₄N₂
MW: 320,18 g/mol
MDL Number: MFCD04973310
CAS číslo: 286014-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR