4-Nitrocinnamaldehyde predominantly trans 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(4-Nitrophenyl)acrylaldehyde
A11467.06
A11467.14
A11467.18
A11467.06EA
2780
CZK
A11467.06
A11467.14
A11467.18
4-Nitrocinnamaldehyde predominantly trans 98%
4-Nitrocinnamaldehyde
Purity: 98%
CAS No.: 1734-79-8
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Odour: Characteristic
Melting Point: 138-142°
Beilstein Reference: 1565424
MDL No.: MFCD00007379
Vzorec:
C₉H₇NO₃ MW: 177,16 g/mol Bod tání: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007379 CAS číslo: 1734-79-8 EINECS: 217-076-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...