3-Methylbenzo[b]thiophene-2-carboxylic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
B24624.03
B24624.06
B24624.14
B24624.03EA
1010
CZK
B24624.03
B24624.06
B24624.14
3-Methylbenzo[b]thiophene-2-carboxylic acid ≥97%
3-Methylbenzo[b]thiophene-2-carboxylic acid
Purity: 97%
CAS No.: 3133-78-6
Molecular Formula: C10H8O2S
Formula Weight: 192.24
Melting Point: 241-244°
MDL No.: MFCD01830305
Vzorec:
C₁₀H₈O₂S MW: 192,24 g/mol Bod tání: 241…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD01830305 CAS číslo: 3133-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...