N-Ethylpyridine-4-methylamine 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine
L13714.14
L13714.06
L13714.14EA
3160
CZK
L13714.14
L13714.06
N-Ethylpyridine-4-methylamine 96%
N-Ethylpyridine-4-methylamine
Purity: 96%
CAS No.: 33403-97-3
Molecular Formula: C8H12N2
Formula Weight: 136.20
Flash Point: 89°(192°F)
Refractive Index: 1.5175
Boiling Point: 103-105°/8mm
Beilstein Reference: 471685
MDL No.: MFCD00023632
Vzorec:
C₈H₁₂N₂ MW: 136,20 g/mol Bod varu: 103…105 °C (8 mmHg) Hustota: 0,98 Bod vzplanutí: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023632 CAS číslo: 33403-97-3 EINECS: 251-506-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...